
(R)-(-)-Prolinol 99%
:68832-13-3
:
:C₅H₁₁NO
:MFCD00064321
:101.1 g/mol
:75 °C (2 mmHg)
:1.02 g/cm³
:GHS07
:Refrigerator
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4830 to 1.4870 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:-29° to -33° (20°C, 589 nm) (c=1, toluene)
Frequently Bought Together