Order Entry
Denmark
ContactUsLinkComponent
3-(Trimethoxysilyl)propyl methacrylate
3-(Trimethoxysilyl)propyl methacrylate
Catalog # SERA28739.01
Supplier:  Serva
CAS Number:  
3-(Trimethoxysilyl)propyl methacrylate
Catalog # SERA28739.01
Supplier:  Serva
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :248.35 g/mol
EINECS:219-785-8
CAS Number:2530-85-0
Formula:C₁₀H₂₀O₅Si
MDL Number:MFCD00008593