Pesticide reference standard, Diazinon 1000 µg/ml in n-nonane
Catalog # CAMBULM-6575-S-10X
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:333-41-5
- Description:Diazinon 1,000 µg/ml in n-nonane
- Formula:C₁₂H₂₁N₂O₃PS
- Description:Diazinon