Sucrose solutions for refractometers
Supplier: Atago
New Product
Certificates
About this item
Sucrose solutions are used for confirming the Brix measurements of refractometers.
- These solutions have a shelf life of 6 weeks
Frequently Bought Together