Order Entry
Denmark
ContactUsLinkComponent
Slide holder, SP Bel-Art
Slide holder, SP Bel-Art
Catalog # 631-9108
undefined
Slide holder, SP Bel-Art
Catalog # 631-9108
Specifications
  • No. of slides:
    10
  • Colour:
    Clear
  • Description:
    Slide holder strip
  • Material:
    PS
  • W×D×H:
    250×50×30 mm
  • Pk:
    1
Specifications
About this item
Slide holder designed to accomodate up to ten slides.
  • Plastic durable construction for regular use
  • Inclined channel to hold slides vertically
  • Front ledge for drying slides