Pesticide reference standard, Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate) 100 µg/ml in n-nonane
Catalog # CAMBULM-8144-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:56-38-2
- Description:Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate) 100 µg/ml in n-nonane
- Formula:C₁₀H₁₄NO₅PS
- Description:Parathion (Parathion-ethyl, O,O-Diethyl O-4-nitrophenyl phosphorothioate)