Silicon standard solution, 1000 mg/L Si in 0,5 M sodium hydroxide solution (from SiO2) Certipur® standard for AAS, Supelco®
Supplier: Merck
Certificates
About this item
Certifications: Traceable to SRM from NIST
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberCertipur
Product Literature
MERC1.70236.0500_TDS_EN
Datasheet
msds_170236