Europium standard solution, 1000 mg/L Eu in 5% nitric acid (from Eu?O?), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Eu₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010992
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00010992
Specifications
Related Information
Frequently Bought Together