Nickel standard solution, 10000 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Ni in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00011137
Specifications
Related Information
Frequently Bought Together