Order Entry
Denmark
ContactUsLinkComponent
Nickel standard solution, 10 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Nickel standard solution, 10 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Catalog # 45261.AE
CAS Number:  
Nickel standard solution, 10 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Catalog # 45261.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00011137
  • Element:
    Nickel
  • Symbol:
    Ni

Specifications

About this item

Molecular Formula: Ni in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137

Certifications: Traceable to NIST