Dysprosiumstandard, 1g/L Dy i 5% salpetersyre (from Dy₂O₃) Specpure® enkeltelement standard til ICP
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Dy₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010982
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00010982
Specifications
Related Information
Frequently Bought Together