Order Entry
Denmark
ContactUsLinkComponent
2-(Propyloxy)ethanol for synthesis, Sigma-Aldrich®
2-(Propyloxy)ethanol for synthesis, Sigma-Aldrich®
Catalog # 8.43947.1000
Supplier:  Merck
CAS Number:  
2-(Propyloxy)ethanol for synthesis, Sigma-Aldrich®
Catalog # 8.43947.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Melting Point:
    -75 °C
  • Vapor Pressure:
    2.1 hPa (20 °C)
  • Flash Point:
    48 °C
  • Density:
    0.912 g/cm3 (20 °C)
  • Ignition Point:
    230°C

Specification Test Results

About this item

Synonyms: Ethylene glycol monopropyl ether, 2-Propoxyethanol, Propyl glycol
 :
 :104.15 g/mol
ADR:3 (6.1),III
 :M-Clarity™ quality level = MQ200
EINECS:220-548-6
UN:1986
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:2807-30-9
Formula:C₅H₁₂O₂
MDL Number:MFCD00072645