Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzene

Benzene

1 Bottle
Catalog # CAMBULM-7039
Flammable
Health
Benzene
Catalog # CAMBULM-7039
CAS Number:  71-43-2
Benzene
Benzene

1 Bottle

CAS Number:  71-43-2
Price
Inquire For Price
Log in for availability
1 * 1 Fl
Product Details & Documents
Synonyms:  Cyclohexatriene
  • CAS Number:71-43-2
  • Formula:C₆H₆
  • MDL Number:MFCD00003009
  • Molecular Weight:78.11 g/mol
  • Merck Index:13,01066
  • Boiling Pt:80.1 °C (1013 hPa)
  • Melting Pt:5.5 °C
  • Density:0.878 g/cm³ (20 °C)
  • Flash Pt:–11 °C
  • Storage Temperature:Ambient
  • EINECS:200-753-7
  • UN:1114
  • ADR:3,II
  • REACH:01-2119447106-44
Specifications

Specifications

  • Pk:
    1 Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.