Order Entry
Denmark
ContactUsLinkComponent
 
Calcium standard solution, 1,000 mg/L Ca in 0.5 M nitric acid standard for AAS
Catalog # REGEAACAH
Supplier:  REAGECON
CAS Number:  
Calcium standard solution, 1,000 mg/L Ca in 0.5 M nitric acid standard for AAS
Catalog # REGEAACAH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item