Order Entry
Denmark
ContactUsLinkComponent
 
Cobalt standard solution, 1000 mg/L Co in 0.5 M nitric acid standard for AAS
Catalog # REGEAACOH
Supplier:  REAGECON
CAS Number:  
Cobalt standard solution, 1000 mg/L Co in 0.5 M nitric acid standard for AAS
Catalog # REGEAACOH
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Cobalt
  • Symbol:
    Co

Specifications

About this item