Ethyl oleate ≥95.0% (by GC)
: TCI
:111-62-6
:85049-36-1
:C₂₀H₃₈O₂
:MFCD00009579
:310.52 g/mol
:14,06828
:209°C
:–32 °C
:0,87 g/cm³ (20 °C)
:181°C
:Ambient
:203-889-5
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together