Order Entry
Denmark
ContactUsLinkComponent
SpinPrep™ PCR clean up kit
SpinPrep™ PCR clean up kit
Catalog # 70976-3
CAS Number:  
SpinPrep™ PCR clean up kit
Catalog # 70976-3
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    SpinPrep™ PCR® Clean-Up Kit
  • Description:
    SpinPrep™ PCR® Clean-Up Kit

Specifications

About this item

The SpinPrep™ PCR Clean Up Kit is designed for the rapid purification of DNA amplified by PCR. The 10-minute procedure involves addition of a binding buffer followed by adsorption of the DNA to a silica membrane in a spin column format. Following a wash step, the DNA is eluted in low salt buffer. This kit removes DNA polymerises, dNTPs, salts, and >99% of primers so that they do not interfere with downstream applications such as cloning, sequencing, or labelling. PCR products from 100 to >12000 bp can be cleaned up, with recoveries of amplified DNA in the range of 60 to 90% under standard conditions.