ABsolute Blue™ qPCR Mixes
Supplier: Thermo Fisher Scientific
Certificates
About this item
ABsolute Blue™ Master Mix incorporates an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy and foolproof. Applications include gene expression, siRNA validation, SNP typing and genotyping.
- Specially formulated to achieve ultimate sensitivity and reproducible qPCR data across all qPCR platforms
- All mixes contain Thermo-Start® hot start Taq polymerase for increased sensitivity, and dTTP for increased reaction efficiency
- Easy to identify mis-aliquoted or skipped wells, particularly when white opaque plates or tubes are used
Storage: Store below 0 °C, preferably at −20 °C, in a constant temperature freezer.
Specifications
- Application:qPCR dye
Specifications
Frequently Bought Together