About this item
Materials for the precise calibration and monitoring of pH instruments.
- Traceable to NIST primary standard and PTB
- Maximum deviation ±0,003 pH units
- Measured in an independent accredited calibration laboratory for the pH value measurement (DKD-K-14301)
11 Options Available Below
Product Details & Documents
Materials for the precise calibration and monitoring of pH instruments.
- Traceable to NIST primary standard and PTB
- Maximum deviation ±0,003 pH units
- Measured in an independent accredited calibration laboratory for the pH value measurement (DKD-K-14301)
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1.01961.0025
- 1.01963.0025
- 1.01965.0025
- 1.01960.0001
- 1.01964.0025
- 1.01962.0001
- 1.07204.0105
- 1.07200.0105
- 1.07202.0105
- 1.07205.0105
- 1.07203.0105
- M-Clarity quality level:
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ300
- MQ300
- MQ300
- MQ300
- MQ300
Specifications
Product Family Options
CertiPUR® certified secondary standard reference material
Product Information
- pH valuePack typePk InfoPkAvailabilityPrice
- 1.01961.0025Potassium tetraoxalate dihydratePotassium tetraoxalate dihydrate1,681 (25°C)Glass bottle for solids1×25 g25 gLog in for availability1 * 25 g1.650,00 krSpecifications:
More
Specifications
Cat. No.1.01961.0025M-Clarity quality levelMQ100 - 1.01963.0025Potassium hydrogen tartratePotassium hydrogen tartrate3,639 (25°C)Glass bottle for solids1×25 g25 gLog in for availability1 * 25 g1.670,00 krSpecifications:
More
Specifications
Cat. No.1.01963.0025M-Clarity quality levelMQ100 - 1.01965.0025Potassium hydrogen phthalatePotassium hydrogen phthalate4,005 (25°C)Glass bottle for solids1×25 g25 gLog in for availability1 * 25 g1.650,00 krSpecifications:
More
Specifications
Cat. No.1.01965.0025M-Clarity quality levelMQ100 - 1.01960.0001Potassium dihydrogen phosphate/ disodium hydrogen phosphate (2×25 g)Potassium dihydrogen phosphate/ disodium hydrogen phosphate (2×25 g)6,863/7,416 (25°C)Glass bottle for solids2×25 g2Log in for availability1 * 2 Items3.310,00 krSpecifications:
More
Specifications
Cat. No.1.01960.0001M-Clarity quality levelMQ100 - 1.01964.0025Disodium tetraborate decahydrateDisodium tetraborate decahydrate9,184 (25°C)Glass bottle for solids1×25 g25 gLog in for availability1 * 25 g1.620,00 krSpecifications:
More
Specifications
Cat. No.1.01964.0025M-Clarity quality levelMQ100 - 1.01962.0001Sodium hydrogen carbonate/sodium carbonateSodium hydrogen carbonate/sodium carbonate10,014 (25°C)Glass bottle for solids2×25 g2Log in for availability1 * 2 Items3.310,00 krSpecifications:
More
Specifications
Cat. No.1.01962.0001M-Clarity quality level
CertiPUR® certified secondary standard reference buffer solution (100 ml plastic bottle)
Product Information
- pH valuePack typePk InfoPkAvailabilityPrice
- 1.07204.0105Potassium tetraoxalate dihydratePotassium tetraoxalate dihydrate1,681 (25°C)Plastic bottle5×100 ml5Log in for availability1 * 5 Items1.730,00 krSpecifications:
More
Specifications
Cat. No.1.07204.0105M-Clarity quality levelMQ300 - 1.07200.0105Potassium hydrogen phthalatePotassium hydrogen phthalate4,005 (25°C)Plastic bottle5×100 ml5Log in for availability1 * 5 Items1.730,00 krSpecifications:
More
Specifications
Cat. No.1.07200.0105M-Clarity quality levelMQ300 - 1.07202.0105Potassium dihydrogen phosphate/ disodium hydrogen phosphatePotassium dihydrogen phosphate/ disodium hydrogen phosphate6,863 (25°C)Plastic bottle5×100 ml5Log in for availability1 * 5 Items1.730,00 krSpecifications:
More
Specifications
Cat. No.1.07202.0105M-Clarity quality levelMQ300 - 1.07205.0105Potassium dihydrogen phosphate/ disodium hydrogen phosphatePotassium dihydrogen phosphate/ disodium hydrogen phosphate7,416 (25°C)Plastic bottle5×100 ml5Log in for availability1 * 5 Items1.730,00 krSpecifications:
More
Specifications
Cat. No.1.07205.0105M-Clarity quality levelMQ300 - 1.07203.0105Disodium tetraborate decahydrateDisodium tetraborate decahydrate9,184 (25°C)Plastic bottle5×100 ml5Log in for availability1 * 5 Items1.730,00 krSpecifications:
More
Specifications
Cat. No.1.07203.0105M-Clarity quality levelMQ300
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



