Order Entry
Denmark
ContactUsLinkComponent
 
Corning® Matrigel® Basement Membrane Matrix, Corning
 :  Corning
    
 
undefined
Corning® Matrigel® Basement Membrane Matrix, Corning
 :  Corning
    
 

 

  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Source:
    Engelbreth-Holm-Swarm mouse tumour
  • Quality control:
    Tested for ability to gel quickly and maintain this form with culture medium for a period of 14 days at 37 °C
    Tested for the ability to promote neurite outgrowth of chick dorsal root ganglia cells
    Tested and found negative for bacteria, fungi, and mycoplasma
    Tested for endotoxin (LAL assay)
  • Storage and stability:
    Stable for at least three months at −20 °C or 12 days at 37 °C

 

Frequently Bought Together