UV lamps, EL series
Supplier: UVP ULTRA VIOLET PRODUCTS
Certificates
About this item
Select from lamps with four, six or eight watts. Models available include 254 nm shortwave, 365 nm longwave, or a combination UV wavelengths, or UV and white light versions.
- Basic configuration offers one UV and one white light tube (use white light when working with materials in a dimly-lit area)
- Twin models have two tubes of the same wavelength
- 2UV™ models use two different wavelengths.
- 3UV™ model, available only in the eight watt lamp, gives three UV wavelengths in one lamp - easily switch among the three wavelengths as the application requires
- Rugged design is excellent for high usage
- Llamps are lightweight for easy holding or place the lamp in the stand or cabinet for hands free operation
- Built-in reflector enhances UV illumination
Frequently Bought Together