Diagnostic bottles, narrow neck, round, PET, with thread, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
PET, clear or amber.
- Provided non-sterile
- Can be sterilized using EtO or gamma radiation
- Lined Poylpropylene screw closures
Ideal for storage and shipment of liquid reagents, buffer solutions,
Ordering information: Closures sold separately.
Packaging: Packaged in shrink wrap modules
Frequently Bought Together