Micro surface stirrer shafts, BOLA
Supplier: Bohlender
Certificates
About this item
Micro stirrer shafts made of PTFE-jacketed stainless steel with blade made completely of PTFE. With four round paddles.
- Resistant to chemicals
- Temperature resistance from –200 to +250 °C
Ideal for stirring in test tubes or narrow-mouth vessels, especially suitable for mixing in round vessels and in vessels with low fill level.
Frequently Bought Together