Storage units, MagLab™ series
Leverandør: HEATHROW
Certifikater
Om denne vare
The MagLab™ series allows common laboratory products to be organised off the benchtop.
ABS, coloured.
- MagRack™ holds four 50 ml tubes and nine 15 ml tubes
- MagWipe™ holds a standard (11×21 cm) Kimwipes® box
- MagPette™ holds two pipettors (fits most popular styles)
- MagBox™ holds miscellaneous accessories
Available individually or as a set of four, each unit is secured by magnets that hold onto most metal surfaces.
Frequently Bought Together