Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® Silicone Fluids
VWR® Silicone Fluids
VWR® Silicone Fluids
Supplier:  VWR Chemicals
Certificates
VWR® Silicone Fluids
VWR® Silicone Fluids
6 Options Available Below
Product Details & Documents
A range of silicone fluids recommended for use in baths. Low in toxicity, inert and long lasting due to their low volatility.
  • Non corrosive
  • High flash point
  • Long service life
  • Low evaporation
Fluids possess excellent heat transfer characteristics and cover a large temperature range from −40 to +200 °C.
1 cS = 1 mm²/s
Packaging: Packed in a plastic bottle/plastic container.
Caution: Harmful vapours may be given off at elevated temperatures. It is advisable to use this material in a fume cupboard at temperatures greater than 140 °C.
Product Family Options
Product Information
  • Temperature range
    Viscosity
    Colour
    Pack type
    Pk
    Availability
    Price
  • 630057W
    Silicone fluid 20 cS
    Silicone fluid 20 cS
    −40...+200 °C
    20 mm²/s
    Clear
    Plastic container
    20 kg
    Log in for availability
    Log in for availability
    1 * 20 kg
    1 * 20 kg - 61.620,00 kr
    61.620,00 kr
    1 * 20 kg
       61.620,00 kr
  • 630076C
    Silicone fluid 100 cS
    Silicone fluid 100 cS
    −40...+200 °C
    100 mm²/s
    Clear
    Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 13.510,00 kr
    13.510,00 kr
    1 * 5 kg
       13.510,00 kr
  • 630077D
    Silicone fluid 100 cS
    Silicone fluid 100 cS
    −40...+200 °C
    100 mm²/s
    Clear
    25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 43.940,00 kr
    43.940,00 kr
    1 * 25 kg
       43.940,00 kr
  • 630094E
    Silicone fluid 350 cS
    Silicone fluid 350 cS
    −40...+200 °C
    350 mm²/s
    Clear
    Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1.810,00 kr
    1.810,00 kr
    1 * 500 g
       1.810,00 kr
  • 630096G
    Silicone fluid 350 cS
    Silicone fluid 350 cS
    −40...+200 °C
    350 mm²/s
    Clear
    Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 14.330,00 kr
    14.330,00 kr
    1 * 5 kg
       14.330,00 kr
  • 630097H
    Silicone fluid 350 cS
    Silicone fluid 350 cS
    −40...+200 °C
    350 mm²/s
    Clear
    Plastic container
    25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 23.800,00 kr
    23.800,00 kr
    1 * 25 kg
       23.800,00 kr
Anbefalinger