Order Entry
Denmark
ContactUsLinkComponent
Methylstyrene (mixture of isomers) stabilised for synthesis, Sigma-Aldrich®
Flammable
Irritant
Methylstyrene (mixture of isomers) stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
Methylstyrene (mixture of isomers) stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    0.09 g/l (20 °C)
  • Melting Point:
    -77 °C
  • Molar Mass:
    118.18 g/mol
  • Boiling Point:
    168 °C
  • Vapor Pressure:
    1.47 hPa (20 °C)
  • Flash Point:
    52 °C
  • Explosion Limit:
    1.1 - 5.2 %(V)
  • Density:
    0.893 g/cm3 (25 °C)
  • Ignition Point:
    575 °C
  • Synonyms:
    Vinyltoluene
  • Hill Formula:
    C9H10
  • Chemical Formula:
    3/4-(CH3)C6H4CH=CH2

Specification Test Results

Related Information

Frequently Bought Together