Order Entry
Denmark
ContactUsLinkComponent
2,4-Diaminophenol dihydrochloride for synthesis, Sigma-Aldrich®
Toxic
Health
2,4-Diaminophenol dihydrochloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Health
2,4-Diaminophenol dihydrochloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    275 g/l (15 °C)
  • Melting Point:
    205 °C
  • Molar Mass:
    197.06 g/mol
  • Bulk Density:
    310 kg/m3
  • pH:
    1 - 2 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together