Calcium thioglycolate trihydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Calcium mercaptoacetate trihydrate, Calcium thioglycolate trihydrate, Calcium bis(mercaptoacetate) trihydrate, Mercaptoacetic acid calcium salt trihydrate
CAS Number:65208-41-5
:MFCD00043437
Formula:C₄H₆CaO₄S₂·3H₂O
MDL Number:MFCD00150707
:276.35 g/mol
Melting Pt:270…280 °C
EINECS:212-402-5
UN:3263
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration Iodometric:≥98.5 %
Specification Test Results
Frequently Bought Together