Order Entry
Denmark
Orders LinkContactUsLinkComponent
Aluminon

Aluminon

Irriterende
Leverandør:   Thermo Fisher Scientific
Aluminon
Certifikater
Aluminon

3 tilgængelige muligheder nedenfor

Product Details & Documents

CAS No.: 569-58-4 Molecular Formula: C22H23N3O9 Formula Weight: 473.44 Odour: Odourless pH: 5.3 Melting Point: 220-225° dec. MDL No.: MFCD00040925

Synonymer:  ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
  • CAS nummer:569-58-4
  • Formel:C₂₂H₂₃N₃O₉
  • MDL nummer:MFCD00040925
  • Molecular Weight:473.44 g/mol
  • Merck Index:14,322
  • Smeltende Pt:220…225 °C (dec.)
  • Opbevaringstemperatur:Ambient
  • EINECS:209-319-1
Product Family Options

Produktinformation

  • Pack type
    Pk
    Tilgængelighed
    Price
  • Glass bottle for solids
    10 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 g
    1 * 10 g - 390,00 kr
    390,00 kr
    1 * 10 g
       390,00 kr

  • Glass bottle for solids
    50 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 g
    1 * 50 g - 1.210,00 kr
    1.210,00 kr
    1 * 50 g
       1.210,00 kr

  • Glass bottle for solids
    250 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 250 g
    1 * 250 g - 4.620,00 kr
    4.620,00 kr
    1 * 250 g
       4.620,00 kr

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.