Aluminon
: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
- :569-58-4
- :C₂₂H₂₃N₃O₉
- :MFCD00040925
- :473.44 g/mol
- :220…225 °C (dec.)
- :Ambient
- :209-319-1
: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
- :569-58-4
- :C₂₂H₂₃N₃O₉
- :MFCD00040925
- :473.44 g/mol
- :220…225 °C (dec.)
- :Ambient
- :209-319-1
- Appearance:Red to yellow-brown to red-brown Crystalline powder
- Infrared spectrum:Conforms
- Sulfated ash:≤0.2 %
- Solubility:(1.0 % in water) Clear
- Suitability:(for Al determination) Passes test
Product Family Options
- Pack typePk



