Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dextran, M.W. ~ 60,000

Dextran, M.W. ~ 60,000

Glass bottle for solids, 500 g
Catalog # SERA18692.02
Marketsource
Supplier:   Serva
Dextran, M.W. ~ 60,000
Catalog # SERA18692.02
Supplier:  Serva
Marketsource
CAS Number:  9004-54-0
Dextran, M.W. ~ 60,000

Glass bottle for solids, 500 g

CAS Number:  9004-54-0
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
  • EINECS:232-677-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications