Order Entry
Denmark
ContactUsLinkComponent
Calcium chloride dihydrate
  RC-030
 :  
Calcium chloride dihydrate
  RC-030
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
 :
 :147.01 g/mol
 :13,01660
 :1600 °C (1013 hPa)
 :176 °C
 :1.85 g/cm³ (20 °C)
 :Ambient
 :233-140-8
 :GHS07
 :10035-04-8
 :CaCl₂·2H₂O
 :MFCD00149613