Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dextran, M.W. ~ 70,000

Dextran, M.W. ~ 70,000

Glass bottle for solids, 500 g
Catalog # A1847.0500
Supplier:   PanReac AppliChem
Dextran, M.W. ~ 70,000
Catalog # A1847.0500
Supplier:  PanReac AppliChem
CAS Number:  9004-54-0
Dextran, M.W. ~ 70,000

Glass bottle for solids, 500 g

CAS Number:  9004-54-0
Price
4.360,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
  • EINECS:232-677-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.