Order Entry
Denmark
ContactUsLinkComponent
Hydroxylammonium chloride, Pierce™
Hydroxylammonium chloride, Pierce™
Catalog # PIER26103
CAS Number:  
undefined
Hydroxylammonium chloride, Pierce™
Catalog # PIER26103
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :69.49 g/mol
Merck Index:13,04853
Boiling Pt:306 °C (1013 hPa)
Melting Pt:151 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-798-2
UN:2923
ADR:8,III
 :GHS05
 :GHS08
 :GHS07
 :GHS09
CAS Number:5470-11-1
Formula:NH₂OH·HCl
MDL Number:MFCD00051089