Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cell spreaders, Delta™

Cell spreaders, Delta™

Cell spreaders, disposable, sterile, 172 mm, individually packed
Catalog # HEATHS8151
Supplier:   HEATHROW
Cell spreaders, Delta™
Catalog # HEATHS8151
Supplier:  HEATHROW
Cell spreaders, Delta™
Cell spreaders, Delta™

Cell spreaders, disposable, sterile, 172 mm, individually packed

Price
1.310,00 kr
Log in for availability
1 * 500 Items
Product Details & Documents

Triangular shaped spreading bar has an edgeless design which prevents tearing of the agar and head shape that distributes pressure evenly for smooth spreading. Handle flexes slightly under pressure and is rippled for easy manipulation. Dimensions 172×37×4 mm.

Documents

Specifications
  • Packed:
    individually packed
  • Description:
    Cell spreaders, disposable, sterile
  • Length:
    172 mm