Politainer
Om denne vare
The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.
- Consists of LDPE and EVA copolymer, which offers optimum protection against volatile solvents
- Available with the frequently used threads GL38 and S60/61 and with volumes of 5, 10 and 20 L
SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces.
Ordering information: The matching boxes with UN-Y approval are available in the table below.
4 tilgængelige muligheder nedenfor
Product Details & Documents
The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.
- Consists of LDPE and EVA copolymer, which offers optimum protection against volatile solvents
- Available with the frequently used threads GL38 and S60/61 and with volumes of 5, 10 and 20 L
SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces.
Ordering information: The matching boxes with UN-Y approval are available in the table below.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
TDS Canisters Overview
Datasheet
Product Family Options
Produktinformation
- DescriptionCapacityColourThreadTypeW×D×HTilgængelighedPrice
- Politainer5 LNaturalGL 38UN-Y approval in combination with box178×178×178 mmLog ind for tilgængelighed1 * 1 Items87,80 kr
- Politainer10 LNaturalGL 38UN-Y approval in combination with box228×228×228 mmLog ind for tilgængelighed1 * 1 Items106,00 kr
- Politainer10 LNaturalS60/61UN-Y approval in combination with box228×228×228 mmLog ind for tilgængelighed1 * 1 Items106,00 kr
- Politainer20 LNaturalS60/61UN-Y approval in combination with box285×285×285 mmLog ind for tilgængelighed1 * 1 Items132,00 kr
Boxes
Produktinformation
- Katalog #W×D×HForDescriptionTilgængelighedPrice
Accessories
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- SCTV107338Thread adapter, PTFE, GL 38 on GL 45Foldable politainer with GL 38Log ind for tilgængelighed1 * 1 Items - 575,00 kr
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



