Desiccator cabinets, SICCO Super-Maxi series
Supplier: Bohlender
Certificates
About this item
Clear, PMMA, reinforced aluminium frame with panels made of acrylic glass.
- Temperature resistance from −20 to +70 °C
- Four casters (two of the casters with brakes)
- Door with magnetic catch and circular rubber seal
- Four removable telescopic shelves made of stainless steel
Desiccators provide ideal conditions for safe and secure storing of poisonous chemicals. Rare or inert gas such as nitrogen can be introduced as soon as the door is closed tightly.
Delivery: Maxi 1-desiccator includes four shelves made of stainless steel, tray, hygrometer and hose (5 m) with two quick couplings for gas filling, four casters, usable with up to 17 shelves. Maxi 2-desiccator includes four shelves made of stainless steel, two trays, two hygrometers and hose (2×5 m) with four quick couplings for gas filling, four casters, usable with up to 8 shelves per compartment.
Frequently Bought Together