Spouts for jerricans
: S.C.A.T.
For simple and safe pouring of fluids from containers.
- Plastic
- Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S55, S60/61, and S40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.
For simple and safe pouring of fluids from containers.
- Plastic
- Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S55, S60/61, and S40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.
Product Family Options
- DescriptionThread
- Flexible spout with safety ventS60/611 * 1 Items132,00 kr
- Rigid spout, electroconductiveS60/611 * 1 Items1.770,00 kr
- Flexible spout with safety ventS70/711 * 1 Items132,00 kr