Bottles, narrow neck, round, HDPE, with screw caps, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
HDPE, natural or white.
- Good chemical resistance
- Narrow mouth ideal for liquids
- Pre-attached white polypropylene screw cap with foamed polyethylene liner
Frequently Bought Together