Order Entry
Denmark
ContactUsLinkComponent
Silicone oil, 500 cSt
Silicone oil, 500 cSt
Catalog # 84539.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Silicone oil, 500 cSt
Catalog # 84539.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Viscosity (25°C):
    450 - 550 cSt
  • Volatile matter:
    Max. 1.0 %

Specification Test Results

About this item

 :
Density:0.97 g/cm³ (20 °C)
Flash Pt:321 °C (closed cup)
Storage Temperature:Ambient
CAS Number:68083-14-7
 :63148-62-9
MDL Number:MFCD00132673