Test strips, sulphite, Reflectoquant®, Supelco®
Katalog # 1.16987.0001
Supplier: Merck
CAS Number:
Specifications
- Chemical method:Nitroprusside / Zn-hexacyanoferrate
- Pk:50 Tests
- Description:Sulphite test
- M-Clarity quality level:MQ100
- Range:10 - 200 mg/l SO₃
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly