Order Entry
Denmark
ContactUsLinkComponent
Test strips, total hardness, Reflectoquant®, Supelco®
Test strips, total hardness, Reflectoquant®, Supelco®
Catalog # 1.16997.0001
Supplier:  Merck
CAS Number:  
undefined
Test strips, total hardness, Reflectoquant®, Supelco®
Catalog # 1.16997.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Phthalein complexone
  • Pk:
    50 Tests
  • Description:
    Total hardness test
  • M-Clarity quality level:
    MQ100
  • Range:
    0,1 - 30,0 °d (1 - 300 mg/l CaO)
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly