Test strips, lead, MQuant®, Supelco®
Lead test, 20-40-100-200-500 mg/l Pb, Rhodizonic acid, MQ, 100 Tests
Supplier: Merck
Jump to:
Product Details & Documents
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: just dip, wait and measure visually against colour scale
- Safe: no auxiliary reagents necessary
- Immediate results within 2 minutes enables fast decisions
- Portable: can be used inside or outside the laboratory
Documents
Specifications
Specifications
- Chemical method:Rhodizonic acid
- Pk:100 Tests
- Description:Lead test
- Type:MQ
- Range:20-40-100-200-500 mg/l Pb
- Cat. No.:1.10077.0001
- M-Clarity quality level:MQ100
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



