Order Entry
Denmark
Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips, Ammonia

QUANTOFIX® Test Strips, Ammonia

QUANTOFIX® aluminum, 0 • 5 • 20 • 50 • 200 • 500 - mg/L - Al³⁺, Color reaction with aurintricarboxylate, 148×122×40 mm (packaging), 100 Tests, 100 Tests
Catalog # 705-0308
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips, Ammonia
Catalog # 705-0308
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips, Ammonia
QUANTOFIX® Test Strips, Ammonia

QUANTOFIX® aluminum, 0 • 5 • 20 • 50 • 200 • 500 - mg/L - Al³⁺, Color reaction with aurintricarboxylate, 148×122×40 mm (packaging), 100 Tests, 100 Tests

CAS Number:  
Price
546,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

These test strips are for the fast and reliable determination of Ammonium, respectively Ammonia, in solutions. The easy dip-and-read procedure provides safe results within 40 seconds. Ammonium/Ammonia is harmful to fish and other aquatic life in aquariums. This test shows the amount of Ammonium quickly and easily.

  • The aquarium can be kept in good condition and fish stay healthy Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

Always ready to use, no accessories necessary for evaluation. Safe results since the desiccant in the stopper for optimal protection of the strips against humidity.

Specifications

Specifications

  • Chemical method:
    Color reaction with aurintricarboxylate
  • Pk:
    100 Tests
  • Description:
    QUANTOFIX® aluminum
  • Dimensions:
    148×122×40 mm (packaging)
  • Tests per kit:
    100 Tests
  • Range:
    0 • 5 • 20 • 50 • 200 • 500 - mg/L - Al³⁺
  • Cat. No.:
    705-0308
  • Parameters:
    Requires additional reagent
  • Contents:
    100 test strips in a tube, additional regaents, instruction leaflet in a box
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    Hazardous material
  • Instrument:
    No
  • Shelf life (months):
    30 (from production)
  • Method:
    Aluminum - Aurin tricarboxylic acid (pink → red)
  • Weight:
    219 g

Specifications