Leather gloves, Grain Driver
Supplier: HONEYWELL SAFETY
About this item
Cowhide grain leather gloves in gunn cut style with an elasticated back.
- Calf grain provides comfort, outstanding flexibility and good mechanical resistance
- Elasticated wrist offers additional support
- Winged thumb and reinforced index finger make the glove more anatomical
The gloves are ideal for general handling in dry environments.
Certifications: EN 388:2016 (3121X); Cat. II
2 Options Available Below
Product Details & Documents
Cowhide grain leather gloves in gunn cut style with an elasticated back.
- Calf grain provides comfort, outstanding flexibility and good mechanical resistance
- Elasticated wrist offers additional support
- Winged thumb and reinforced index finger make the glove more anatomical
The gloves are ideal for general handling in dry environments.
Certifications: EN 388:2016 (3121X); Cat. II
Specifications for Product Family
Specifications
- Catalog No:
- 112-1456
- 112-1455
- Model:
- Driver
- Driver
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Cuff style:
- Elastic
- Elastic
- EN norm:
- EN 388:2016 (3121X)
- EN 388:2016 (3121X)
Specifications
Product Family Options
Product Information
- Glove sizeColourLengthThicknessAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.112-1456ModelDriverAmbidextrous / hand-specificHand-specificDisposable / reusableReusableCuff styleElasticEN normEN 388:2016 (3121X) - Specifications:
More
Specifications
Cat. No.112-1455ModelDriverAmbidextrous / hand-specificHand-specificDisposable / reusableReusableCuff styleElasticEN normEN 388:2016 (3121X)