Organic reference standard, Bromochloroacetic acid 1000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERYA10713000MB
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:5589-96-8
- Description:Bromochloroacetic acid 1,000 µg/ml in tert-butyl methyl ether
- Formula:C₂H₂BrClO₂
- Description:Bromochloroacetic acid