Order Entry
Denmark
ContactUsLinkComponent
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Flammable
Corrosive
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7142.1000
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Corrosive
Perchloric acid 0.1 mol/l (0.1 N) in anhydrous acetic acid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7142.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (mol/l):
    0.098 - 0.102

Specification Test Results

About this item

 :
 :100.46 g/mol
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8,II
 :GHS02
 :GHS05
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325