Order Entry
Denmark
ContactUsLinkComponent
Celite® Hyflo Super-cel®, filter aid
Health
Celite® Hyflo Super-cel®, filter aid
Catalog # ROTH0003.1
Supplier:  Roth Carl
CAS Number:  
Health
Celite® Hyflo Super-cel®, filter aid
Catalog # ROTH0003.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

About this item

 :
Boiling Pt:2200 °C (1013 hPa)
Melting Pt:1710 °C
Density:2.3 g/cm³ (20 °C)
 :GHS08
CAS Number:68855-54-9
MDL Number:MFCD00132803