Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 1,4-Dichlorobutane 2000 µg/ml in methanol
Organic reference standard, 1,4-Dichlorobutane 2000 µg/ml in methanol
Supplier:  Restek
Certificates
Organic reference standard, 1,4-Dichlorobutane 2000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    110-56-5
  • Formula:
    C₄H₈Cl₂
  • Description:
    1,4-Dichlorobutane

Specifications

Frequently Bought Together