Order Entry
Denmark
ContactUsLinkComponent
 
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
 
undefined
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™

 

  • Assay (GC):
    Min. 99.5 %
  • Boiling range:
    114 - 116 °C
  • Explosion Limit:
    12.4 %
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Ammonia (NH₃):
    Max. 0.001 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000003 %
  • Iron (Fe):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • Chloride (Cl):
    Max. 0.0005 %
  • Sulfate (SO₄):
    Max. 0.0005 %
  • KMnO₄ reducing matter (as O):
    Max. 0.0005 %
  • Piperidine (GC):
    Max. 0.01 %
  • 2-picoline (GC):
    Max. 0.2 %
  • Solubility in water:
    complying
  • Refractive Index:
    n20/D 1.509 (lit.)

 

 

Frequently Bought Together